2,3-Dichloro-N-[2-(2,6-dimethylphenoxy)propyl]aniline

Supplier: Matrix Scientific

031684-500MG
101832-194EA 306.15 USD
101832-194
2,3-Dichloro-N-[2-(2,6-dimethylphenoxy)propyl]aniline
2,3-Dichloro-N-[2-(2,6-dimethylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MW: 324.25 g/mol
MDL Number: MFCD10687731
CAS Number: 1040682-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR