N-(2-Butoxybenzyl)-2,3-dichloroaniline

Supplier: Matrix Scientific

031683-500MG
101832-192EA 306.15 USD
101832-192
N-(2-Butoxybenzyl)-2,3-dichloroaniline
N-(2-Butoxybenzyl)-2,3-dichloroaniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MDL Number: MFCD10687730
CAS Number: 1040682-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR