N-(4-Butoxybenzyl)-2,3-dichloroaniline

Supplier: Matrix Scientific

031682-500MG
101832-190EA 306.15 USD
101832-190
N-(4-Butoxybenzyl)-2,3-dichloroaniline
N-(4-Butoxybenzyl)-2,3-dichloroaniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MDL Number: MFCD10687729
CAS Number: 1036512-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR