2,3-Dichloro-N-(2-ethoxybenzyl)aniline
Supplier: Matrix Scientific
031680-500MG
101832-186EA
306.15
USD
101832-186
2,3-Dichloro-N-(2-ethoxybenzyl)aniline
2,3-Dichloro-N-(2-ethoxybenzyl)aniline
Hazard: Irritant
Formula:
C₁₅H₁₅Cl₂NO |
MDL Number:
MFCD10687727 CAS Number: 1036584-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...