2,3-Dichloro-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

031675-500MG
101832-176EA 306.15 USD
101832-176
2,3-Dichloro-N-(3-methylbenzyl)aniline
2,3-Dichloro-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₃Cl₂N
MDL Number: MFCD10687723
CAS Number: 1040042-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR