2,3-Dichloro-N-[2-(2-isopropylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031674-500MG
101832-174EA 306.15 USD
101832-174
2,3-Dichloro-N-[2-(2-isopropylphenoxy)ethyl]aniline
2,3-Dichloro-N-[2-(2-isopropylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MW: 324.25 g/mol
MDL Number: MFCD10687722
CAS Number: 1040681-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR