3-Chloro-N-(4-ethoxybenzyl)-4-methylaniline

Supplier: Matrix Scientific

031672-500MG
101832-170EA 306.15 USD
101832-170
3-Chloro-N-(4-ethoxybenzyl)-4-methylaniline
3-Chloro-N-(4-ethoxybenzyl)-4-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₈ClNO
MDL Number: MFCD10687720
CAS Number: 1036537-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR