3-Chloro-N-[2-(2,5-dimethylphenoxy)propyl]-4-methylaniline

Supplier: Matrix Scientific

031671-500MG
101832-168EA 306.15 USD
101832-168
3-Chloro-N-[2-(2,5-dimethylphenoxy)propyl]-4-methylaniline
3-Chloro-N-[2-(2,5-dimethylphenoxy)propyl]-4-methylaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD10687719
CAS Number: 1040681-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR