3-Chloro-N-[2-(4-methoxyphenoxy)ethyl]-4-methylaniline

Supplier: Matrix Scientific

031667-500MG
101832-160EA 306.15 USD
101832-160
3-Chloro-N-[2-(4-methoxyphenoxy)ethyl]-4-methylaniline
3-Chloro-N-[2-(4-methoxyphenoxy)ethyl]-4-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₈ClNO₂
MDL Number: MFCD10687715
CAS Number: 1040681-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR