2,4,5-Trichloro-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline

Supplier: Matrix Scientific

031666-500MG
101832-158EA 306.15 USD
101832-158
2,4,5-Trichloro-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline
2,4,5-Trichloro-N-[4-(tetrahydro-2-furanylmethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₁₈H₁₈Cl₃NO₂
MDL Number: MFCD10687714
CAS Number: 1040681-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR