2,4,5-Trichloro-N-(2-ethoxybenzyl)aniline

Supplier: Matrix Scientific

031664-500MG
101832-154EA 306.15 USD
101832-154
2,4,5-Trichloro-N-(2-ethoxybenzyl)aniline
2,4,5-Trichloro-N-(2-ethoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₄Cl₃NO
MDL Number: MFCD10687712
CAS Number: 1036533-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR