N-[3-(2-Phenoxyethoxy)benzyl]cyclohexanamine

Supplier: Matrix Scientific

031658-500MG
101832-142EA 306.15 USD
101832-142
N-[3-(2-Phenoxyethoxy)benzyl]cyclohexanamine
N-[3-(2-Phenoxyethoxy)benzyl]cyclohexanamine

Hazard: Irritant

Formula: C₂₁H₂₇NO₂
MDL Number: MFCD10687707
CAS Number: 1040680-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR