N-(4-Chlorophenethyl)cyclohexanamine

Supplier: Matrix Scientific

031654-500MG
101832-134EA 306.15 USD
101832-134
N-(4-Chlorophenethyl)cyclohexanamine
N-(4-Chlorophenethyl)cyclohexanamine

Hazard: Irritant

Formula: C₁₄H₂₀ClN
MDL Number: MFCD10687703
CAS Number: 855635-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR