N-[2-(3,4-Dimethylphenoxy)ethyl]cyclohexanamine

Supplier: Matrix Scientific

031653-500MG
101832-132EA 306.15 USD
101832-132
N-[2-(3,4-Dimethylphenoxy)ethyl]cyclohexanamine
N-[2-(3,4-Dimethylphenoxy)ethyl]cyclohexanamine

Hazard: Irritant

Formula: C₁₆H₂₅NO
MDL Number: MFCD10687702
CAS Number: 1040680-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR