[2-(2-Ethoxyethoxy)phenyl]-N-(tetrahydro-2-furanylmethyl)methanamine

Supplier: Matrix Scientific

031648-500MG
101832-122EA 306.15 USD
101832-122
[2-(2-Ethoxyethoxy)phenyl]-N-(tetrahydro-2-furanylmethyl)methanamine
[2-(2-Ethoxyethoxy)phenyl]-N-(tetrahydro-2-furanylmethyl)methanamine

Hazard: Irritant

Formula: C₁₆H₂₅NO₃
MDL Number: MFCD10687699
CAS Number: 1040680-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR