(3-Ethoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine

Supplier: Matrix Scientific

031646-500MG
101832-118EA 306.15 USD
101832-118
(3-Ethoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine
(3-Ethoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD07188566
CAS Number: 880804-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR