(2-Butoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine

Supplier: Matrix Scientific

031643-500MG
101832-112EA 306.15 USD
101832-112
(2-Butoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine
(2-Butoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine

Hazard: Irritant

Formula: C₁₆H₂₅NO₂
MDL Number: MFCD10687695
CAS Number: 1040692-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR