2-(4-Ethylphenoxy)-N-(tetrahydro-2-furanylmethyl)-1-ethanamine

Supplier: Matrix Scientific

031640-500MG
101832-106EA 306.15 USD
101832-106
2-(4-Ethylphenoxy)-N-(tetrahydro-2-furanylmethyl)-1-ethanamine
2-(4-Ethylphenoxy)-N-(tetrahydro-2-furanylmethyl)-1-ethanamine

Hazard: Irritant

Formula: C₁₅H₂₃NO₂
MDL Number: MFCD10687692
CAS Number: 1040692-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR