N-(4-Isobutoxybenzyl)-1-butanamine

Supplier: Matrix Scientific

031634-500MG
101832-094EA 306.15 USD
101832-094
N-(4-Isobutoxybenzyl)-1-butanamine
N-(4-Isobutoxybenzyl)-1-butanamine

Hazard: Irritant

Formula: C₁₅H₂₅NO
MDL Number: MFCD10687687
CAS Number: 1040691-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR