N-[4-(Hexyloxy)benzyl]-1-butanamine

Supplier: Matrix Scientific

031633-500MG
101832-092EA 306.15 USD
101832-092
N-[4-(Hexyloxy)benzyl]-1-butanamine
N-[4-(Hexyloxy)benzyl]-1-butanamine

Hazard: Irritant

Formula: C₁₇H₂₉NO
MDL Number: MFCD10687686
CAS Number: 1040691-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR