N-[2-(4-Ethylphenoxy)ethyl]-1-butanamine

Supplier: Matrix Scientific

031632-500MG
101832-090EA 306.15 USD
101832-090
N-[2-(4-Ethylphenoxy)ethyl]-1-butanamine
N-[2-(4-Ethylphenoxy)ethyl]-1-butanamine

Hazard: Irritant

Formula: C₁₄H₂₃NO
MDL Number: MFCD02633045
CAS Number: 420101-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR