N-[4-(tert-Butyl)benzyl]-1-butanamine

Supplier: Matrix Scientific

031631-500MG
101832-088EA 306.15 USD
101832-088
N-[4-(tert-Butyl)benzyl]-1-butanamine
N-[4-(tert-Butyl)benzyl]-1-butanamine

Hazard: Irritant

Formula: C₁₅H₂₅N
MDL Number: MFCD10687685
CAS Number: 875305-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR