N-Butyl-4-(2,4-dichlorophenoxy)-1-butanamine

Supplier: Matrix Scientific

031630-500MG
101832-086EA 306.15 USD
101832-086
N-Butyl-4-(2,4-dichlorophenoxy)-1-butanamine
N-Butyl-4-(2,4-dichlorophenoxy)-1-butanamine

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD01309687
CAS Number: 418784-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR