N-[2-(4-Chlorophenoxy)ethyl]-1-butanamine

Supplier: Matrix Scientific

031628-500MG
101832-082EA 306.15 USD
101832-082
N-[2-(4-Chlorophenoxy)ethyl]-1-butanamine
N-[2-(4-Chlorophenoxy)ethyl]-1-butanamine

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD02633086
CAS Number: 67746-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR