N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-1-butanamine

Supplier: Matrix Scientific

031626-500MG
101832-078EA 306.15 USD
101832-078
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-1-butanamine
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-1-butanamine

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD02106305
CAS Number: 420100-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR