N-(3,5-Dichlorobenzyl)-1-heptanamine

Supplier: Matrix Scientific

031624-500MG
101832-074EA 306.15 USD
101832-074
N-(3,5-Dichlorobenzyl)-1-heptanamine
N-(3,5-Dichlorobenzyl)-1-heptanamine

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂N
MDL Number: MFCD10687683
CAS Number: 90390-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR