N-[2-(2-Ethoxyethoxy)benzyl]-1-heptanamine

Supplier: Matrix Scientific

031623-500MG
101832-072EA 306.15 USD
101832-072
N-[2-(2-Ethoxyethoxy)benzyl]-1-heptanamine
N-[2-(2-Ethoxyethoxy)benzyl]-1-heptanamine

Hazard: Irritant

Formula: C₁₈H₃₁NO₂
MDL Number: MFCD10687682
CAS Number: 1040691-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR