N-(3-Propoxybenzyl)-1-heptanamine

Supplier: Matrix Scientific

031622-500MG
101832-070EA 306.15 USD
101832-070
N-(3-Propoxybenzyl)-1-heptanamine
N-(3-Propoxybenzyl)-1-heptanamine

Hazard: Irritant

Formula: C₁₇H₂₉NO
MDL Number: MFCD10687681
CAS Number: 1040691-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR