N-(2-Fluorobenzyl)-1-heptanamine

Supplier: Matrix Scientific

031621-500MG
101832-068EA 306.15 USD
101832-068
N-(2-Fluorobenzyl)-1-heptanamine
N-(2-Fluorobenzyl)-1-heptanamine

Hazard: Irritant

Formula: C₁₄H₂₂FN
MDL Number: MFCD07407697
CAS Number: 893589-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR