N-[2-(4-Chloro-2-methylphenoxy)ethyl]-1-heptanamine

Supplier: Matrix Scientific

031618-500MG
101832-062EA 306.15 USD
101832-062
N-[2-(4-Chloro-2-methylphenoxy)ethyl]-1-heptanamine
N-[2-(4-Chloro-2-methylphenoxy)ethyl]-1-heptanamine

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD03211101
CAS Number: 138323-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR