N-[2-(2-Ethoxyethoxy)benzyl]-3-methoxyaniline

Supplier: Matrix Scientific

031616-500MG
101832-058EA 306.15 USD
101832-058
N-[2-(2-Ethoxyethoxy)benzyl]-3-methoxyaniline
N-[2-(2-Ethoxyethoxy)benzyl]-3-methoxyaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₃
MDL Number: MFCD10687677
CAS Number: 1040691-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR