N-[2-(4-Isopropylphenoxy)propyl]-3-methoxyaniline
Supplier: Matrix Scientific
031613-500MG
101832-052EA
306.15
USD
101832-052
N-[2-(4-Isopropylphenoxy)propyl]-3-methoxyaniline
N-[2-(4-Isopropylphenoxy)propyl]-3-methoxyaniline
Hazard: Irritant
Formula:
C₁₉H₂₅NO₂ |
MDL Number:
MFCD10687674 CAS Number: 1040691-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...