3-Methoxy-N-(4-propoxybenzyl)aniline

Supplier: Matrix Scientific

031612-500MG
101832-050EA 306.15 USD
101832-050
3-Methoxy-N-(4-propoxybenzyl)aniline
3-Methoxy-N-(4-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁NO₂
MDL Number: MFCD10687673
CAS Number: 1036543-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR