N-(3-Methoxyphenyl)-N-[4-(pentyloxy)benzyl]amine

Supplier: Matrix Scientific

031611-500MG
101832-048EA 306.15 USD
101832-048
N-(3-Methoxyphenyl)-N-[4-(pentyloxy)benzyl]amine
N-(3-Methoxyphenyl)-N-[4-(pentyloxy)benzyl]amine

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687672
CAS Number: 1040690-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR