3-Methoxy-N-neopentylaniline

Supplier: Matrix Scientific

031610-500MG
101832-046EA 306.15 USD
101832-046
3-Methoxy-N-neopentylaniline
3-Methoxy-N-neopentylaniline

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD10687671
CAS Number: 1040690-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR