3-Methoxy-N-[2-(2-methoxyphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031609-500MG
101832-044EA 306.15 USD
101832-044
3-Methoxy-N-[2-(2-methoxyphenoxy)ethyl]aniline
3-Methoxy-N-[2-(2-methoxyphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₉NO₃
MDL Number: MFCD10687670
CAS Number: 1040690-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR