N-[2-(2,4-Dichlorophenoxy)propyl]-3-methoxyaniline

Supplier: Matrix Scientific

031608-500MG
101832-042EA 306.15 USD
101832-042
N-[2-(2,4-Dichlorophenoxy)propyl]-3-methoxyaniline
N-[2-(2,4-Dichlorophenoxy)propyl]-3-methoxyaniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO₂
MDL Number: MFCD03211201
CAS Number: 356538-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR