3-Methoxy-N-[2-(4-methylphenoxy)ethyl]aniline
Supplier: Matrix Scientific
031607-500MG
101832-040EA
306.15
USD
101832-040
3-Methoxy-N-[2-(4-methylphenoxy)ethyl]aniline
3-Methoxy-N-[2-(4-methylphenoxy)ethyl]aniline
Hazard: Irritant
Formula:
C₁₆H₁₉NO₂ |
MDL Number:
MFCD10687669 CAS Number: 1040690-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...