N-(3-Isobutoxybenzyl)-1-phenyl-1-ethanamine

Supplier: Matrix Scientific

031605-500MG
101832-036EA 306.15 USD
101832-036
N-(3-Isobutoxybenzyl)-1-phenyl-1-ethanamine
N-(3-Isobutoxybenzyl)-1-phenyl-1-ethanamine

Hazard: Irritant

Formula: C₁₉H₂₅NO
MDL Number: MFCD10687668
CAS Number: 1040690-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR