2,4-Dichloro-N-[2-(4-methoxyphenoxy)propyl]aniline

Supplier: Matrix Scientific

031604-500MG
101832-034EA 306.15 USD
101832-034
2,4-Dichloro-N-[2-(4-methoxyphenoxy)propyl]aniline
2,4-Dichloro-N-[2-(4-methoxyphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO₂
MDL Number: MFCD10687667
CAS Number: 1040690-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR