N-(2-Furylmethyl)-1-phenyl-1-ethanamine

Supplier: Matrix Scientific

031597-500MG
101832-020EA 306.15 USD
101832-020
N-(2-Furylmethyl)-1-phenyl-1-ethanamine
N-(2-Furylmethyl)-1-phenyl-1-ethanamine

Hazard: Irritant

Formula: C₁₃H₁₅NO
MDL Number: MFCD03425022
CAS Number: 161119-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR