1-Phenyl-N-(2-thienylmethyl)-1-ethanamine

Supplier: Matrix Scientific

031596-500MG
101832-018EA 306.15 USD
101832-018
1-Phenyl-N-(2-thienylmethyl)-1-ethanamine
1-Phenyl-N-(2-thienylmethyl)-1-ethanamine

Hazard: Irritant

Formula: C₁₃H₁₅NS
MDL Number: MFCD03425023
CAS Number: 680185-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR