N-[2-(3,5-Dimethylphenoxy)ethyl]-1-phenyl-1-ethanamine

Supplier: Matrix Scientific

031594-500MG
101832-014EA 306.15 USD
101832-014
N-[2-(3,5-Dimethylphenoxy)ethyl]-1-phenyl-1-ethanamine
N-[2-(3,5-Dimethylphenoxy)ethyl]-1-phenyl-1-ethanamine

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687660
CAS Number: 1040690-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR