2-Methyl-N-(2-propoxybenzyl)-2-propanamine
Supplier: Matrix Scientific
031591-500MG
101832-008EA
306.15
USD
101832-008
2-Methyl-N-(2-propoxybenzyl)-2-propanamine
2-Methyl-N-(2-propoxybenzyl)-2-propanamine
Hazard: Irritant
Formula:
C₁₄H₂₃NO |
MDL Number:
MFCD06197182 CAS Number: 866816-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...