N-[2-(2,4-Dichlorophenoxy)ethyl]-2-methyl-2-propanamine

Supplier: Matrix Scientific

031586-500MG
101831-998EA 306.15 USD
101831-998
N-[2-(2,4-Dichlorophenoxy)ethyl]-2-methyl-2-propanamine
N-[2-(2,4-Dichlorophenoxy)ethyl]-2-methyl-2-propanamine

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD01827020
CAS Number: 356532-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR