N-(tert-Butyl)-3-phenyl-1-propanamine

Supplier: Matrix Scientific

031585-500MG
101831-996EA 306.15 USD
101831-996
N-(tert-Butyl)-3-phenyl-1-propanamine
N-(tert-Butyl)-3-phenyl-1-propanamine

Hazard: Irritant

Formula: C₁₃H₂₁N
MDL Number: MFCD03210878
CAS Number: 71193-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR