N-[2-(2-Isopropylphenoxy)ethyl]-2-methyl-2-propanamine

Supplier: Matrix Scientific

031583-500MG
101831-992EA 306.15 USD
101831-992
N-[2-(2-Isopropylphenoxy)ethyl]-2-methyl-2-propanamine
N-[2-(2-Isopropylphenoxy)ethyl]-2-methyl-2-propanamine

Hazard: Irritant

Formula: C₁₅H₂₅NO
MDL Number: MFCD10687652
CAS Number: 1040689-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR