N-Benzyl-3-(2-chlorophenyl)-1-propanamine

Supplier: Matrix Scientific

031581-500MG
101831-988EA 306.15 USD
101831-988
N-Benzyl-3-(2-chlorophenyl)-1-propanamine
N-Benzyl-3-(2-chlorophenyl)-1-propanamine

Hazard: Irritant

Formula: C₁₆H₁₈ClN
MDL Number: MFCD10687651
CAS Number: 566913-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR