N-Benzyl-2-(3-methylphenoxy)-1-butanamine

Supplier: Matrix Scientific

031577-500MG
101831-980EA 306.15 USD
101831-980
N-Benzyl-2-(3-methylphenoxy)-1-butanamine
N-Benzyl-2-(3-methylphenoxy)-1-butanamine

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687647
CAS Number: 1040689-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR