N-Benzyl-2-(2,6-dimethylphenoxy)-1-ethanamine

Supplier: Matrix Scientific

031576-500MG
101831-978EA 306.15 USD
101831-978
N-Benzyl-2-(2,6-dimethylphenoxy)-1-ethanamine
N-Benzyl-2-(2,6-dimethylphenoxy)-1-ethanamine

Hazard: Irritant

Formula: C₁₇H₂₁NO
MDL Number: MFCD10687646
CAS Number: 1040689-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR